  • Search Results
  • Do combined alternating sessions of 1540 nm nonablative fractional laser and percutaneous collagen induction with trichloroacetic acid 20% show better results than each individual modality in the treatment of atrophic acne scars? A randomized controlled trial.
Do combined alternating sessions of 1540 nm nonablative fractional laser and percutaneous collagen induction with trichloroacetic acid 20% show better results than each individual modality in the treatment of atrophic acne scars? A randomized controlled trial.
The Journal of dermatological treatment (2012-05-30)
Tahra M Leheta, Rania M Abdel Hay, Rehab A Hegazy, Yehia F El Garem
ABSTRACT
There have been no well-controlled studies evaluating the efficacy of combining 1540 nm nonablative fractional laser with percutaneous collagen induction (PCI) and trichloroacetic acid (TCA) 20% in the treatment of atrophic acne scars. We hypothesized that combined alternating sessions of both modalities would show better results than each individual modality. Thirty-nine patients with post acne atrophic scars were included in this study. Patients were randomly equally divided into three groups; group 1 was subjected to six sessions of PCI combined with TCA 20% in the same session, group 2 was subjected to six sessions of 1540 nm fractional laser and group 3 was subjected to combined alternating sessions of the previously mentioned two modalities. Scar severity scores improved by a mean of 59.79% (95% CI 47.38-72.21) (p < 0.001) in group 1, a mean of 61.83% (95% CI 54.09-69.56) (p < 0.001) in group 2 and a mean of 78.27% (95% CI 74.39-82.15) (p < 0.001) in group 3. The difference in the degree of improvement was statistically significant when comparing the three groups using ANOVA test (p = 0.004). The current work recommends combining 1540 nm nonablative fractional laser in alternation with PCI and TCA 20% in the treatment of atrophic acne scars.
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%