  • Search Results
  • In vivo induction of cutaneous inflammation results in the accumulation of extracellular trap-forming neutrophils expressing RORγt and IL-17.
In vivo induction of cutaneous inflammation results in the accumulation of extracellular trap-forming neutrophils expressing RORγt and IL-17.
The Journal of investigative dermatology (2013-12-10)
Romy R M C Keijsers, Anke G M Hendriks, Piet E J van Erp, Bram van Cranenbroek, Peter C M van de Kerkhof, Hans J P M Koenen, Irma Joosten
ABSTRACT
Clinical trials successfully using antibodies targeting IL-17 in psoriasis support the importance of IL-17 in the pathophysiology of this disease. However, there is a debate concerning the source and dynamics of IL-17 production in inflamed skin. Here we characterized IL-17-producing immune cells over time, using two established in vivo models of human skin inflammation that share many histological features with psoriasis, i.e., leukotriene B4 application and tape-stripping. Both treatments revealed a clear influx of neutrophils and T cells. Staining for IL-17 revealed that the majority of IL-17 was expressed by neutrophils and mast cells, in both models. Neutrophils, but not mast cells, coexpressed the IL-17-associated transcription factor RORγt and were able to form extracellular traps. While the presence of mast cells remained steady during the skin inflammatory process, the presence of neutrophils was clearly dynamic in time. Therefore, it is attractive to hypothesize that IL-17+/RORγt+ neutrophils contribute to human skin inflammation in vivo and possibly to the pathogenesis of skin diseases such as psoriasis. Surprisingly, T cells represented a minority of the IL-17-expressing cell population. These observations challenge the classical opinion that IL-17 is predominantly associated with T cells in skin inflammation.
Sigma-Aldrich
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.