  • Search Results
  • Transdifferentiation of differentiated ovary into functional testis by long-term treatment of aromatase inhibitor in Nile tilapia.
Transdifferentiation of differentiated ovary into functional testis by long-term treatment of aromatase inhibitor in Nile tilapia.
Endocrinology (2014-01-21)
Li-Na Sun, Xiao-Long Jiang, Qing-Ping Xie, Jing Yuan, Bao-Feng Huang, Wen-Jing Tao, Lin-Yan Zhou, Yoshitaka Nagahama, De-Shou Wang
ABSTRACT
Females with differentiated ovary of a gonochoristic fish, Nile tilapia, were masculinized by long-term treatment with an aromatase inhibitor (Fadrozole) in the present study. The reversed gonads developed into functional testes with fertile sperm. The longer the fish experienced sex differentiation, the longer treatment time was needed for successful sex reversal. Furthermore, Fadrozole-induced sex reversal, designated as secondary sex reversal (SSR), was successfully rescued by supplement of exogenous 17β-estradiol. Gonadal histology, immunohistochemistry, transcriptome, and serum steroid level were analyzed during SSR. The results indicated that spermatogonia were transformed from oogonia or germline stem cell-like cells distributed in germinal epithelium, whereas Leydig and Sertoli cells probably came from the interstitial cells and granulosa cells of the ovarian tissue, respectively. The transdifferentiation of somatic cells, as indicated by the appearance of doublesex- and Mab-3-related transcription factor 1 (pre-Sertoli cells) and cytochrome P450, family 11, subfamily B, polypeptide 2 (pre-Leydig cells)-positive cells in the ovary, provided microniche for the transdifferentiation of germ cells. Decrease of serum 17β-estradiol was detected earlier than increase of serum 11-ketotestosterone, indicating that decrease of estrogen was the cause, whereas increase of androgen was the consequence of SSR. The sex-reversed gonad displayed more similarity in morphology and histology with a testis, whereas the global gene expression profiles remained closer to the female control. Detailed analysis indicated that transdifferentiation was driven by suppression of female pathway genes and activation of male pathway genes. In short, SSR provides a good model for study of sex reversal in teleosts and for understanding of sex determination and differentiation in nonmammalian vertebrates.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
11-Ketotestosterone
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fadrozole hydrochloride, ≥98% (HPLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard