  • Search Results
  • Strategies to prevent the neuropsychiatric side-effects of corticosteroids: a case report and review of the literature.
Strategies to prevent the neuropsychiatric side-effects of corticosteroids: a case report and review of the literature.
Current opinion in organ transplantation (2014-02-21)
Stephen West, Chris Kenedi
ABSTRACT
The use of corticosteroids is increasing, and while the physical complications of their use are well known, the neuropsychiatric consequences are not. This review focuses on preventing these neuropsychiatric complications. Although there are limited data on this subject, it is a problem that clinicians face on a regular basis. The incidence of neuropsychiatric complications rises rapidly once the daily dose of prednisone is greater than 40 mg. Other risk factors for neuropsychiatric symptoms are damaged blood-brain barrier and hypoalbuminemia. All patients receiving corticosteroids and their caregivers should be warned about the potential neuropsychiatric complications. Small trials have supported the use of various agents as prophylaxis. The development of neuropsychiatric symptoms secondary to corticosteroids should lead to prompt involvement of liaison psychiatry. There is a lack of large randomized controlled studies to inform clinical practice. At present, lithium and olanzapine probably represent the best choices for prophylaxis. Patients with a prior history of steroid-related psychosis or mania should be considered for prophylaxis when future courses of steroids are prescribed as limited data, and our clinical experience suggests that this can reduce the future episodes of neuropsychiatric side-effects.
Sigma-Aldrich
(±)-Lorazepam
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Lorazepam, European Pharmacopoeia (EP) Reference Standard
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Supelco
Lorazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Olanzapine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Prednisone, ≥98%
USP
Gabapentin, United States Pharmacopeia (USP) Reference Standard
USP
Olanzapine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gabapentin, solid
Supelco
6α-Methylprednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Supelco
Gabapentin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gabapentin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Methylprednisolone, United States Pharmacopeia (USP) Reference Standard
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard
Olanzapine, European Pharmacopoeia (EP) Reference Standard
Supelco
Gabapentin Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material
Olanzapine for system suitability, European Pharmacopoeia (EP) Reference Standard