  • Search Results
  • Alkali burn versus suture-induced corneal neovascularization in C57BL/6 mice: an overview of two common animal models of corneal neovascularization.
Alkali burn versus suture-induced corneal neovascularization in C57BL/6 mice: an overview of two common animal models of corneal neovascularization.
Experimental eye research (2014-02-25)
Chiara Giacomini, Giulio Ferrari, Fabio Bignami, Paolo Rama
ABSTRACT
The purpose of the present study was to quantify and compare corneal hem- and lymphangiogenesis between alkali burn and suture-induced corneal neovascularization (CNV) in two commonly used mouse strains. A retrospective analysis was performed on C57BL/6 and FVB neovascularized corneas. CNV was induced by surface caustication with NaOH or intrastromal placement of three 10.0 nylon sutures. Hemangiogenesis and lymphangiogenesis extent was calculated on whole mounted corneas by CD31 and LYVE1 immunofluorescence analysis. Blood vessel growth was similar between alkali burn and suture-induced CNV in C57BL/6 mice, and between C57BL/6 and FVB sutured strains. On the contrary, corneal lymphangiogenesis was more pronounced in the C57BL/6 sutured mice versus the alkali burn group, and in the FVB strain versus both C57BL/6 models. These results indicate that significant differences occur in lymphangiogenesis, but not hemangiogenesis, in the alkali burn and suture-induced models in C57BL/6 mice. Furthermore, lymphangiogenesis is more pronounced in the albino (FVB) strain after suture placement. We suggest that the suture model has a number of advantages and may be preferentially used to study corneal lymphangiogenesis.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh