  • Search Results
  • Calcium/calmodulin-dependent kinases are involved in growth, thermotolerance, oxidative stress survival, and fertility in Neurospora crassa.
Calcium/calmodulin-dependent kinases are involved in growth, thermotolerance, oxidative stress survival, and fertility in Neurospora crassa.
Archives of microbiology (2014-02-27)
Ravi Kumar, Ranjan Tamuli
ABSTRACT
Calcium/calmodulin-dependent kinases (Ca(2+)/CaMKs) are Ser/Thr protein kinases that respond to change in cytosolic free Ca(2+) ([Ca(2+)]c) and play multiple cellular roles in organisms ranging from fungi to humans. In the filamentous fungus Neurospora crassa, four Ca(2+)/CaM-dependent kinases, Ca(2+)/CaMK-1 to 4, are encoded by the genes NCU09123, NCU02283, NCU06177, and NCU09212, respectively. We found that camk-1 and camk-2 are essential for full fertility in N. crassa. The survival of ∆camk-2 mutant was increased in induced thermotolerance and oxidative stress conditions. In addition, the ∆camk-1 ∆camk-2, ∆camk-4 ∆camk-2, and ∆camk-3 ∆camk-2 double mutants display slow growth phenotype, reduced aerial hyphae, decreased thermotolerance, and increased sensitivity to oxidative stress, revealing the genetic interactions among these kinases. Therefore, Ca(2+)/CaMKs are involved in growth, thermotolerance, oxidative stress tolerance, and fertility in N. crassa.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis