  • Search Results
  • Late onset of a persistent, deep stromal scarring after PRK and corneal cross-linking in a patient with forme fruste keratoconus.
Late onset of a persistent, deep stromal scarring after PRK and corneal cross-linking in a patient with forme fruste keratoconus.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-04-08)
Jose L Güell, Paula Verdaguer, Daniel Elies, Oscar Gris, Felicidad Manero
ABSTRACT
To present a case of a late, deep stromal scar in a 22-year-old patient with forme fruste keratoconus who underwent combined corneal cross-linking and photorefractive keratectomy (PRK). Topography-guided corneal cross-linking combined with corneal PRK (without complications) was performed in both eyes with a delay of 2 weeks between each eye. At the 5-month postoperative examination of the right eye, a localized corneal haze was circumscribed to the posterior deep stroma, signifying a decrease of visual acuity. However, this improved partially and temporarily when treated with topical corticoids during 2 years of follow-up and then reoccurred, affecting the corrected distance visual acuity. To the authors' knowledge, this is the first documented, clinical case presenting a deep stromal affectation without endothelial decompensation and visual acuity affectation as a postoperative complication following topography-guided PRK and corneal cross-linking.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Riboflavin (B2), analytical standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.