  • Search Results
  • Gene expression regulation of Bcl2, Bax and cytochrome-C by geraniol on chronic MPTP/probenecid induced C57BL/6 mice model of Parkinson's disease.
Gene expression regulation of Bcl2, Bax and cytochrome-C by geraniol on chronic MPTP/probenecid induced C57BL/6 mice model of Parkinson's disease.
Chemico-biological interactions (2014-04-29)
Karamkolly R Rekha, Govindasamy P Selvakumar
ABSTRACT
Parkinson's disease (PD) is a common disabling movement disorder owing to progressive depletion of dopamine in nigrostriatal region, and can be experimentally accelerated by the neurotoxin 1-methyl-4-phenyl-1, 2, 3, 6-tetrahydropyridine (MPTP). MPTP-treated mice are a representative animal model for searching for the therapeutic agents for PD without adverse effect. In this study we investigated the effect of geraniol (GE) on chronic MPTP/probenecid (MPTP/p) induced apoptotic changes in nigrostriatal region. We observed that chronic exposure to MPTP/p led to increased expression of apoptotic markers, results in neurodegeneration and motor behavioral impairments in mice. Pretreatment with GE to MPTP/p significantly improved motor functions and ameliorated striatal antioxidant balance. In addition, GE attenuated the expression of apoptotic markers evident by the normalized Bcl-2/Bax ratio and decreased expression of cytochrome-C and caspase-9 in the substantia nigra and striatum of MPTP/p induced mice model of PD. The findings of the present study suggested that GE, a new therapeutic potential avenue may have beneficial effects in slowing or preventing the progression of PD and other neurodegenerative disorders.
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, suitable for GFC marker, BioReagent
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Probenecid
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE)
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Supelco
Nerol, analytical standard
USP
Probenecid, United States Pharmacopeia (USP) Reference Standard
Probenecid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Supelco
Geraniol, analytical standard