Metastatic involvement of the maxillary antrum from an uncommon source.
Dental update (2014-05-21)
Rokhsareh Elledge, Ross O C Elledge, Diana Raskauskiene
ABSTRACT
Metastatic involvement of the jawbones is uncommon, particularly in the maxilla. Case reports of such metastases from renal cell primaries are few, making a consensus on treatment difficult to establish. We present a case of metastatic involvement of the maxilla two years following a nephrectomy for renal cell carcinoma. The case exemplifies the broad range of symptoms attributable to metastases in the maxilla and the management dilemmas. The case highlights the role of practitioners in primary dental care in identifying the potential for such pathology based on clinical and radiographic features.
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard