  • Search Results
  • Ancient DNA reveals elephant birds and kiwi are sister taxa and clarifies ratite bird evolution.
Ancient DNA reveals elephant birds and kiwi are sister taxa and clarifies ratite bird evolution.
Science (New York, N.Y.) (2014-05-24)
Kieren J Mitchell, Bastien Llamas, Julien Soubrier, Nicolas J Rawlence, Trevor H Worthy, Jamie Wood, Michael S Y Lee, Alan Cooper
ABSTRACT
The evolution of the ratite birds has been widely attributed to vicariant speciation, driven by the Cretaceous breakup of the supercontinent Gondwana. The early isolation of Africa and Madagascar implies that the ostrich and extinct Madagascan elephant birds (Aepyornithidae) should be the oldest ratite lineages. We sequenced the mitochondrial genomes of two elephant birds and performed phylogenetic analyses, which revealed that these birds are the closest relatives of the New Zealand kiwi and are distant from the basal ratite lineage of ostriches. This unexpected result strongly contradicts continental vicariance and instead supports flighted dispersal in all major ratite lineages. We suggest that convergence toward gigantism and flightlessness was facilitated by early Tertiary expansion into the diurnal herbivory niche after the extinction of the dinosaurs.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta