Surgical treatment of single gingival recessions: clinical guidelines.
European journal of oral implantology (2014-06-04)
Giovanpaolo Pini-Prato, Michele Nieri, Umberto Pagliaro, Teresa Schifter Giorgi, Michele La Marca, Debora Franceschi, Jacopo Buti, Monica Giani, Julia Hanne Weiss, Luigi Padeletti, Pierpaolo Cortellini, Leandro Chambrone, Luca Barzagli, Efisio Defraia, Roberto Rotundo
ABSTRACT
The purpose of this clinical guidelines project was to determine the most appropriate surgical techniques, in terms of efficacy, complications, and patient opinions, for the treatment of buccal single gingival recessions without loss of interproximal soft and hard tissues. Literature searches were performed (electronically and manually) for entries up to 28 February, 2013 concerning the surgical approaches for the treatment of gingival recessions. Systematic reviews (SRs) of randomised controlled trials (RCTs) and individual RCTs that reported at least 6 months of follow-up of surgical treatment of single gingival recessions were included. The full texts of the selected SRs and RCTs were analysed using checklists for qualitative evaluation according to the Scottish Intercollegiate Guidelines Network (SIGN) method. The following variables were evaluated: Complete Root Coverage (CRC); Recession Reduction (RecRed); complications; functional and aesthetic satisfaction of the patients; and costs of therapies. Out of 30 systematic reviews, 3 SRs and 16 out of 313 RCTs were judged to have a low risk for bias (SIGN code: 1+). At a short-term evaluation, the coronally advanced flap plus connective tissue graft method (CAF+CTG) resulted in the best treatment in terms of CRC and/or RecRed; in case of cervical abrasion and presence of root sensitivity CAF + CTG + Restoration caused less sensitivity than CAF+CTG. CAF produced less postoperative discomfort for patients. Limited information is available regarding postoperative dental hypersensitivity and aesthetic satisfaction of the patients. In presence of aesthetic demands or tooth hypersensitivity, the best way to surgically treat single gingival recessions without loss of interproximal tissues is achieved using the CAF procedure associated with CTG. Considering postoperative discomfort, the CAF procedure is the less painful surgical approach, while the level of aesthetic satisfaction resulted higher after CAF either alone or with CTG. It is unclear how much tooth hypersensitivity is reduced by surgically covering buccal recessions. It is important to note that the present recommendations are based on short-term data (less than 1 year). The guidelines project was made possible through self-financing by the authors.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.