  • Search Results
  • Studying SIRT6 regulation using H3K56 based substrate and small molecules.
Studying SIRT6 regulation using H3K56 based substrate and small molecules.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-07-09)
Piia Kokkonen, Minna Rahnasto-Rilla, Paolo Mellini, Elina Jarho, Maija Lahtela-Kakkonen, Tarja Kokkola
ABSTRACT
SIRT6 is a modulator of chromatin structure having an important role in healthy ageing, and there is a crucial need to find specific modulators for it. Therefore, the activity of SIRT6 should be studied using a variety of methods. We examined the capability of SIRT6 to deacetylate a set of five fluorogenic substrates based on p53 and histone H3 sequences. The substrate designed around H3K56 deacetylation site exhibited the best signal-to-background ratio and was chosen for further studies. Nicotinamide is a known inhibitor for sirtuins, and it was found to be less potent inhibitor for SIRT6 than it is for SIRT1. In addition, we studied 15 other small molecule sirtuin modulators using the H3K56 based substrate. EX-527, quercetin and three pseudopeptidic compounds were found to be the most potent SIRT6 inhibitors, exhibiting over 50% deacetylation inhibition. These findings describe the first modulators of SIRT6 activity at the physiologically important H3K56 deacetylation site.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material