MilliporeSigma
Mechanisms of L-cysteine neurotoxicity.
Neurochemical research (2000-11-04)
R Janáky, V Varga, A Hermann, P Saransaari, S S Oja
ABSTRACT
We review here the possible mechanisms of neuronal degeneration caused by L-cysteine, an odd excitotoxin. L-Cysteine lacks the omega carboxyl group required for excitotoxic actions via excitatory amino acid receptors, yet it evokes N-methyl-D-aspartate (NMDA) -like excitotoxic neuronal death and potentiates the Ca2+ influx evoked by NMDA. Both actions are prevented by NMDA antagonists. One target for cysteine effects is thus the NMDA receptor. The following mechanisms are discussed now: (1) possible increase in extracellular glutamate via release or inhibition of uptake/degradation, (2) generation of cysteine alpha-carbamate, a toxic analog of NMDA, (3) generation of toxic oxidized cysteine derivatives, (4) chelation of Zn2+ which blocks the NMDA receptor-ionophore, (5) direct interaction with the NMDA receptor redox site(s), (6) generation of free radicals, and (7) formation of S-nitrosocysteine. In addition to these, we describe another new alternative for cytotoxicity: (8) generation of the neurotoxic catecholamine derivative, 5-S-cysteinyl-3,4-dihydroxyphenylacetate (cysdopac).
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%