When betamethasone and dexamethasone are unavailable: hydrocortisone.
Journal of perinatology : official journal of the California Perinatal Association (2002-03-16)
L E Moore, J N Martin
ABSTRACT
To review the literature on alternatives to betamethasone and dexamethasone for enhancement of fetal lung maturity. A medline search was conducted from 1966 to the present. Trials dealing with enhancement of fetal lung maturity using modalities other than betamethasone or dexamethasone were reviewed. Eight studies met inclusion criteria. Excluding betamethasone and dexamethasone, the most frequently studied glucocorticoids, for fetal lung maturation, are methylprednisolone and hydrocortisone. Methylprednisolone does not cross the placenta. Two grams of hydrocortisone has been shown to improve indices of fetal lung maturity (i.e., L/S ratio) and to improve fetal outcomes compared to no treatment. There is limited information about alternatives to betamethasone and dexamethasone for the enhancement of fetal lung maturity in women at risk of preterm delivery. In the absence of these two preferred drugs, hydrocortisone can be given at a dose of 500 mg intravenously every 12 hours for four doses for this indication.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Betamethasone, ≥98%
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Sigma-Aldrich
Betamethasone, meets USP testing specifications