  • Search Results
  • Photodynamic therapy with verteporfin: a new treatment in ophthalmology.
Photodynamic therapy with verteporfin: a new treatment in ophthalmology.
Seminars in ophthalmology (2004-10-30)
S Michels, U Schmidt-Erfurth
ABSTRACT
Photodynamic therapy (PDT) with verteporfin is a new treatment modality in ophthalmology that has previously shown its effectiveness in treatment of a variety of neoplastic pathologies. In this therapeutic approach, the photosensitizer verteporfin is activated by non-thermal laser light to obtain closure of neovascular structures. Preclinical and clinical studies have indicated that PDT is a safe, selective, and effective treatment for choroidal neovascularization in age-related macular degeneration. No significant damage to the neurosensory retina was found, which explains why PDT does not cause loss of visual acuity and may be used in a larger population than laser photocoagulation. This review summarizes the mechanisms of action of PDT, and the results of preclinical and clinical studies in ophthalmology.
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)