  • Search Results
  • Medicinal chemistry and biological properties of non-imidazole histamine H3 antagonists.
Medicinal chemistry and biological properties of non-imidazole histamine H3 antagonists.
Mini reviews in medicinal chemistry (2004-11-17)
Marlon Cowart, Robert Altenbach, Lawrence Black, Ramin Faghih, Chen Zhao, Arthur A Hancock
ABSTRACT
The H3 receptor is prominently expressed in neuronal tissues, and H3 antagonists have been proposed as drugs with benefits in disorders of cognition, attention, pain, allergic rhinitis, and obesity. The structure-activity relationships (SAR) of various classes of non-imidazole H3 antagonists are reviewed, along with highlights of functional efficacy in tissue-based and animal disease models.
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard