  • Search Results
  • Mechanism of inhibition of DNA methyltransferases by cytidine analogs in cancer therapy.
Mechanism of inhibition of DNA methyltransferases by cytidine analogs in cancer therapy.
Cancer biology & therapy (2004-11-13)
Humaira Gowher, Albert Jeltsch
ABSTRACT
Hypermethylation of tumor suppressor genes caused by aberrant activity of DNA methyltransferases is an important mechanism that contributes to cancer. The reaction mechanism of DNA methyltransferases, which includes formation of a covalent intermediate between the enzyme and the target base, is the basis of the success of several anti-cancer drugs that are targeted against DNA methylation. These include 5-fluoro-2'-deoxycytidine, 5-aza-2'-deoxycytidine (Decitabine) and 2-H pyrimidinone-1-beta-D(2'-deoxyriboside) (Zebularine). This review provides an insight to how the chemistry of DNA methylation is involved in the performance of these drugs targeted against it.
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%