  • Search Results
  • Neurogenesis mediated by gamma-aminobutyric acid and glutamate signaling.
Neurogenesis mediated by gamma-aminobutyric acid and glutamate signaling.
Journal of pharmacological sciences (2009-06-02)
Noritaka Nakamichi, Takeshi Takarada, Yukio Yoneda
ABSTRACT
In this review, we will summarize our ongoing studies on the functionality of both gamma-aminobutyric acid (GABA) and glutamate receptors expressed by undifferentiated neural progenitor cells isolated from embryonic rodent brains. Cells were cultured with growth factors for the formation of round spheres by clustered cells under floating conditions, whereas a reverse transcription polymerase chain reaction analysis revealed expression of mRNA for particular subtypes of different ionotropic and metabotropic GABA and glutamate receptors in undifferentiated progenitors and neurospheres. Moreover, sustained exposure to either GABAergic or glutamatergic agonists not only modulated the size of neurospheres formed, but also affected spontaneous and induced differentiation of neural progenitor cells into particular progeny cell lineages such as neurons and astroglia. Both GABA and glutamate could play a pivotal role in the mechanisms underlying proliferation for self-replication along with the determination of subsequent differentiation fate toward particular progeny lineages through activation of their receptor subtypes functionally expressed by undifferentiated neural progenitor cells. Accordingly, neurogenesis seems to be also under control by GABAergic and glutamatergic signaling in developing brains as seen with neurotransmission in adult brains.
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard