MilliporeSigma
Oral potassium iodide for the treatment of sporotrichosis.
The Cochrane database of systematic reviews (2009-10-13)
Siliang Xue, Rui Gu, Taixiang Wu, Mingming Zhang, Xiaoshan Wang
ABSTRACT
Sporotrichosis is a subacute or chronic disease, usually affecting the skin caused by a dimorphic (existing in two forms), aerobic (oxygen requiring) fungus called Sporothrix schenckii. Oral potassium iodide is widely used for cutaneous sporotrichosis in clinical medicine with more and more reports published. However, the benefits and adverse reactions of these treatments have not yet been systematically reviewed. To assess the effects of oral potassium iodide for the treatment of sporotrichosis. In July 2009 we searched the Cochrane Skin Group Specialised Skin Register, the Cochrane Central Register of Controlled Clinical Trials (CENTRAL) in The Cochrane Library (Issue 3, 2009), MEDLINE and EMBASE, The Chinese Biomedical Database, CNKI, VIP, and ongoing trials registers. Randomised trials comparing orally administered iodide with placebo, or with another treatment. Studies about potassium iodide as an adjunct were excluded. Two authors planned to independently assess trial quality and extract data. We also planned to collect adverse effects information from the trials where possible. In the absence of any suitable randomised placebo-controlled trials or comparisons with other treatments in this area, we were unable to assess the effects of oral potassium iodide. The currently available evidence is insufficient to assess the potential for oral potassium iodide in the treatment of sporotrichosis.There is no high-quality evidence for or against oral potassium iodide as a treatment for sporotrichosis. Further randomised double-blind placebo-controlled trials are needed to define the efficacy and acceptability of these interventions.
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Gold etchant, standard