  • Search Results
  • The unsolved cyclosporine-induced kidney injury: is paricalcitol a feasible new renoprotective option?
The unsolved cyclosporine-induced kidney injury: is paricalcitol a feasible new renoprotective option?
Kidney international (2010-05-29)
Flávio N F Reis
ABSTRACT
The management of cyclosporine A (CsA)-induced nephrotoxicity remains one of the main challenges in kidney transplantation. The animal study by Park et al. proposes that paricalcitol, a vitamin D analog with renoprotective actions reported in other conditions, attenuates CsA-induced kidney injury via the suppression of inflammatory, fibrotic, and apoptotic factors. Before paricalcitol can be considered a feasible new therapeutic option for post-transplantation nephropathy, these interesting data require further studies assessing other mechanisms of CsA-induced nephrotoxicity.
Product Description
USP
Paricalcitol, United States Pharmacopeia (USP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.