  • Search Results
  • The role of carbamazepine and oxcarbazepine in alcohol withdrawal syndrome.
The role of carbamazepine and oxcarbazepine in alcohol withdrawal syndrome.
Journal of clinical pharmacy and therapeutics (2010-05-12)
R Barrons, N Roberts
ABSTRACT
The goal of this review is to evaluate the efficacy and safety of carbamazepine and oxcarbazepine in treatment of alcohol withdrawal syndrome (AWS) and determine the role in therapy of both agents. Relevant literature was identified through a search of MEDLINE (1966-June 2008), PubMed (1966-June 2008); Cochrane database was performed to identify English-language publications. Search terms included carbamazepine, oxcarbazepine, AWS, alcoholism, substance syndrome withdrawal. In seven studies, including 612 patients, carbamazepine demonstrated significant reduction in alcohol withdrawal scores. However, in comparative trials with a benzodiazepine agent, carbamazepine's ability to prevent alcohol withdrawal seizures (OR = 0.93; 95% CI = 0.06-14.97, P = NS) and delirium tremens (DTs; OR = 1.25; 95% CI = 0.28-5.64, P = NS) was uncertain as a result of insufficient patient enrollment. In three trials, carbamazepine failed to reduce alcohol withdrawal symptoms possibly as a result of delayed administration, inadequate dosage or inadequate sample size. At daily doses of 800 mg either fixed or tapered over 5-9 days, carbamazepine was well tolerated, and safely administered when blood alcohol concentration dropped below 0.15%. The role of oxcarbazepine in AWS is undefined because of inconsistent findings in two trials. Carbamazepine has demonstrated safety, tolerability and efficacy in treatment of moderate to severe symptoms of alcohol withdrawal in the inpatient setting. However, trials of carbamazepine provide inconclusive evidence for prevention of alcohol withdrawal seizures and DTs in comparison with benzodiazepines. Benzodiazepines remain the primary treatment of moderate to severe AWS.
Product Description
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Oxcarbazepine, European Pharmacopoeia (EP) Reference Standard
USP
Oxcarbazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid
Supelco
Oxcarbazepine, analytical standard
Supelco
Oxcarbazepine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®