  • Search Results
  • Case report: High chromium and cobalt levels in a pregnant patient with bilateral metal-on-metal hip arthroplasties.
Case report: High chromium and cobalt levels in a pregnant patient with bilateral metal-on-metal hip arthroplasties.
Clinical orthopaedics and related research (2012-06-14)
Juliane Fritzsche, Cornelia Borisch, Christof Schaefer
ABSTRACT
Metal-on-metal bearings frequently are used in young patients leading to the concern that disseminated metals such as chromium (Cr) and cobalt (Co) as the main constituents could affect pregnancies. We describe a 41-year-old patient with bilateral metal-on-metal hip arthroplasties, a recurrent pseudotumor, and extremely high blood levels (Cr 39 μg/L, Co 138 μg/L) at 12 gestational weeks. At different gestational weeks, maternal blood, aspirate of the pseudotumor, and amniotic fluid were analyzed for Cr and Co. Therapy with chelating agents was not recommended because the mother showed no symptoms of toxicity and the safety of chelating therapy during pregnancy is not established. At 38 weeks of gestation, a healthy male infant was delivered with elevated Cr and Co cord blood levels. At the age of 8 weeks, the infant's Cr was comparable to the cord blood level, whereas the Co decreased considerably without treatment. At the age of 14 weeks, the infant's development was seemingly uneventful and no signs of toxicity were obvious. Carcinogenic, mutagenic, and teratogenic potentials of these metals have been suggested. However, we found no published clinical observations in context with pregnancies of women with hip arthroplasties using metal-on-metal implants. To our knowledge, this is the first report of such high levels of Cr and Co in a human pregnancy. Although we cannot generalize from one case, the seemingly uneventful outcome of this pregnancy may reassure colleagues when counseling patients with high ion levels whether to carry a pregnancy to term.
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Cobalt, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Cobalt, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%
Cobalt, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%