  • Search Results
  • [Stenosing and perforated ulcers of the small intestine related to potassium chloride in enteric-coated tablets. Apropos of 11 cases].
[Stenosing and perforated ulcers of the small intestine related to potassium chloride in enteric-coated tablets. Apropos of 11 cases].
Chirurgie; memoires de l'Academie de chirurgie (1989-01-01)
J Grosdidier, P Boissel, L Bresler, A Vidrequin
ABSTRACT
The enteric coated KCl tablet formulation generates complications that affect selectively the small intestine. A novel 1 g KCl tablet preparation proved to be accountable for the development of a specific type of intestinal disease. 11 cases of stenotic ulcers of the proximal ileum have been treated at the Clinique chirurgicale "C" (CHRU, Nancy) since January 1986, 9 of them presenting with perforations. These 11 patients required potassium replacement, which was being supplied by the same tablet formula. A study of these 11 cases focusing on the patients history and clinical characteristics was carried out, that included both macroscopic and microscopic examinations of the lesions. Arterial and venous networks were visualized by X-ray in two surgical tissue specimens, respectively, following injection of contrast material. The authors were thus able to bring forth evidence of the clinical uniqueness (prodromal pain reminiscent of König's syndrome and peritonitis characterized by asthenia); to identify the main risk factor (i.e., reduced bowel movement); as well as describe the basic lesion (pylorus-like stenosis), and propound a novel theory concerning the etiology and the pathology of this condition. The observation of the aforementioned cases initiated a nation-wide, post-marketing pharmacological survey, which resulted, as did a similar investigation which had been carried out in the States, in the discontinuation of any treatment by this type of galenic form of the product.
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%