A global safety evaluation of etodolac.
Clinical rheumatology (1989-03-01)
A Karbowski
ABSTRACT
Etodolac has previously been reported to have an excellent safety profile. Results of recent clinical trials and of a clinical practice study were compared with the previously reported safety data for etodolac. The total incidence of patient complaints and of laboratory abnormalities continues to be low, with no apparent increase in side effects with increasing patient age. In the clinical practice study, 60% of study events occurred early in therapy (before 10 weeks). These data support an excellent safety profile for etodolac.
Sigma-Aldrich
Etodolac
Etodolac for peak identification, European Pharmacopoeia (EP) Reference Standard
Etodolac, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.