MilliporeSigma
Hydroxycobalamin/sodium thiosulfate as a cyanide antidote.
The Journal of emergency medicine (1987-01-01)
A H Hall, B H Rumack
ABSTRACT
Severe, acute cyanide poisoning is uncommon and can be very difficult to diagnose if a history of exposure is unavailable. Victims of smoke inhalation may have significant cyanide poisoning as well as carbon monoxide toxicity. The Lilly Cyanide Antidote Kit currently available in America unfortunately has its own inherent toxicity. An efficacious antidote lacking toxicity is desirable, especially in cases where the diagnosis of cyanide poisoning cannot be made with certainty. Hydroxycobalamin/sodium thiosulfate has been used in France since 1970. Both components have been shown to be safe and efficacious in animal studies. Case reports of human cyanide poisoning treated with hydroxycobalamin/sodium thiosulfate have been published only in French. Animal and human data on the use of this antidotal combination are reviewed. Hydroxycobalamin/sodium thiosulfate is an efficacious cyanide antidote with little inherent toxicity.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Sodium thiosulfate solution, 2 g/dL in deionized water