MilliporeSigma
The methenamine misunderstanding in the therapy of phosgene poisoning.
Archives of toxicology (1980-12-01)
W F Diller
ABSTRACT
A thorough analysis of the pertinent literature leads to the conclusion that there is no justification for the use of methenamine (hexamethylene tetramin, Urotropin) for the therapy of phosgene poisoning.
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
Methenamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Hexamethylenetetramine
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene