  • Search Results
  • Toxicity and metabolism of the neurotoxic hexacarbons n-hexane, 2-hexanone, and 2,5-hexanedione.
Toxicity and metabolism of the neurotoxic hexacarbons n-hexane, 2-hexanone, and 2,5-hexanedione.
Annual review of pharmacology and toxicology (1982-01-01)
D Couri, M Milks
ABSTRACT
Human exposure to hexacarbon compounds is quite pervasive, including occupational exposures to industrial solvents as well as unintentional and sometimes deliberate exposures to hexacarbon solvents contained in innumerable commercial products. The exact mechanism of hexacarbon neurotoxicity has not yet been identified, but an interference with neuronal axoplasmic flow seems most likely. Metabolism of n-hexane and 2-hexanone to 2,5-hexanedione is a prominent feature which appears to be causally related to the neuropathologic syndrome, and mixed solvent effects have been noted in regard to potentiation of hexacarbon neurotoxicity. Continued effort in investigating the chemically induced peripheral neuropathy is essential not only to define the precise molecular mechanism, but to advance our basic understanding of other polyneuropathies as well. Ultimately, progress in these areas should yield such benefits as early diagnosis of potential neuropathology, better measures for the prevention of neurotoxicities, and more effective modalities of treatment. Indeed, sustained research efforts are imperative in maintaining human health and safety throughout our current era of advancing global technology.
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Supelco
Hexane, analytical standard
Sigma-Aldrich
2,5-Hexanedione, ≥98%
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Supelco
2-Hexanone, analytical standard
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hexane, suitable for HPLC, ≥95%
Supelco
2,5-Hexanedione, analytical standard
Supelco
2-Hexanone, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.