  • Search Results
  • MR arthrography: impact of steroids, local anesthetics, and iodinated contrast material on gadolinium signal intensity in phantoms at 1.5 and 3.0 T.
MR arthrography: impact of steroids, local anesthetics, and iodinated contrast material on gadolinium signal intensity in phantoms at 1.5 and 3.0 T.
Radiology (2014-03-26)
Marco A Ugas, Bang H Huynh, Michael G Fox, James T Patrie, Cree M Gaskin
ABSTRACT
To determine if gadolinium signal intensity at direct magnetic resonance (MR) arthrography is affected by the addition of steroids, anesthetics, or iodinated contrast material. This study did not require approval by the institutional review board because no patients or patient data were involved. An in vitro study was performed to evaluate various concentrations of three gadolinium contrast agents (gadopentetate dimeglumine, gadobenate dimeglumine, and gadofosveset trisodium) diluted in either saline or iodinated contrast material (50% and full-strength iohexol 300). Three steroids (betamethasone, triamcinolone, and methylprednisolone) and three local anesthetics (lidocaine, ropivacaine, and bupivacaine) were added to solutions in clinical doses. T1-weighted fat-suppressed MR imaging sequences were performed in phantoms at 1.5 and 3.0 T. Signal intensities were measured. All experiments were repeated in full for a total of three replications each. The data were analyzed by using two-way factorial analysis of variance. Dilution of gadolinium into iohexol reduced the signal intensity in all samples compared with dilution in saline alone. Peak signal intensities were at 0.625 and 1.25 mmol/L of gadolinium in iohexol at both magnet strengths. At 1.5 T, the addition of steroids and anesthetics to the saline solutions had no impact on the signal intensity curves, with the peak signal intensity at gadolinium concentrations of 2.5 and 1.25 mmol/L. At 3.0 T, the addition of steroids and anesthetics had minimal effect on signal intensity curves, with the peak signal intensity at 1.25 mmol/L of gadolinium. The addition of steroids and/or anesthetics to gadolinium solutions for MR arthrography does not substantially impact signal intensity. When gadolinium is diluted into a 50% or greater strength of iohexol, the signal intensity curve shifts so that the maximum signal intensity is obtained with lower gadolinium concentrations (0.625-1.25 mmol/L).
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Lidocaine, powder
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)
Sigma-Aldrich
Meglumine, meets USP testing specifications
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Supelco
Iohexol, analytical standard
USP
Betamethasone, United States Pharmacopeia (USP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Gadolinium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Gadolinium, chips
Supelco
Betamethasone, VETRANAL®, analytical standard
USP
Methylprednisolone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gadolinium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)