Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, 2m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.0125mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 50x50mm, thickness 6.0mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 50mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.008mm, 99.99+%