  • Search Results
  • Percutaneous vascular closure using an anchored collagen plug provides effective haemostasis following both antegrade and retrograde femoral arterial punctures.
Percutaneous vascular closure using an anchored collagen plug provides effective haemostasis following both antegrade and retrograde femoral arterial punctures.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2014-06-01)
A Chaudhuri, A York, R Dey
ABSTRACT
Small published series suggest a higher failure rate for Angio-Seal vascular closure device (VCD) deployment after antegrade femoral puncture, despite the need for shorter haemostasis times, early discharge, and possibly higher turnover. We seek to compare the deployment efficacy and complications of the Angio-Seal VCD between antegrade and retrograde femoral arterial deployments. Radiological data was retrospectively analysed from prospective databases from the hospitals' Computerised Radiology Information System (CRIS) over 2010-2012. Angio-Seal gauge, Rutherford class (as applicable), puncture mode (used to classify deployment as antegrade/retrograde), sheath sizes, and deployment success/failures were recorded. Numerical/statistical analyses were undertaken using Microsoft Excel 10/SISA software. A total of 519 Angio-Seal VIP VCDs were deployed in 470 patients over 2010-2012 (13 other patients could not be analysed due to incomplete data). Sheath sizes for antegrade/retrograde femoral puncture were 5F, n = 22/9; 6F, n = 244/223; 7F, n = 1/5; 9F, n = 4/0. 8F Angio-Seal VIPs were used for 9F punctures only, 6F for the remainder. The overall deployment success rate was 93.7%. In total, 247 (91.1%) successful antegrade deployments were undertaken with 24 (8.9%) failures, compared with 229 (96.6%) successful retrograde deployments with eight (3.4%) failures. Antegrade/retrograde failures were classed as failure to deploy, n = 15/5; bleeding despite successful deployment requiring supplementary compression, n = 6/1; haematoma formation, n = 2/1; groin pain, n = 0/1; vessel stenosis, n = 1/0. Higher deployment failures were noted with antegrade deployment (p < .02, chi-square test). Angio-Seal deployment is successful for both antegrade/retrograde femoral punctures albeit with a higher antegrade failure rate.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.