  • Search Results
  • Differences that matter: major cytotoxic T cell-stimulating minor histocompatibility antigens.
Differences that matter: major cytotoxic T cell-stimulating minor histocompatibility antigens.
Immunity (2000-10-06)
S Malarkannan, T Horng, P Eden, F Gonzalez, P Shih, N Brouwenstijn, H Klinge, G Christianson, D Roopenian, N Shastri
ABSTRACT
Despite thousands of genetic polymorphisms among MHC matched mouse strains, a few unknown histocompatibility antigens are targeted by the cytotoxic T cells specific for tissue grafts. We isolated the cDNA of a novel BALB.B antigen gene that defines the polymorphic H28 locus on chromosome 3 and yields the naturally processed ILENFPRL (IFL8) peptide for presentation by Kb MHC to C57BI/6 CTL. The CTL specific for the IFL8/Kb and our previously identified H60/Kb complexes represent a major fraction of the B6 anti-BALB.B immune response. The immunodominance of these antigens can be explained by their differential transcription in the donor versus the host strains and their expression in professional donor antigen-presenting cells.