  • Search Results
  • Oleic Acid, deglycosylated vitamin D-binding protein, nitric oxide: a molecular triad made lethal to cancer.
Oleic Acid, deglycosylated vitamin D-binding protein, nitric oxide: a molecular triad made lethal to cancer.
Anticancer research (2014-07-02)
Marco Ruggiero, Emma Ward, Rodney Smith, Jacopo J V Branca, David Noakes, Gabriele Morucci, Margit Taubmann, Lynda Thyer, Stefania Pacini
ABSTRACT
Oleic Acid (OA) has been shown to have anticancer properties mediated by interaction with proteins such as α-lactalbumin and lactoferrins. Therefore, we synthesized complexes of OA and Gc protein-derived macrophage activating factor (GcMAF) that inhibits per se cancer cell proliferation and metastatic potential. We hypothesised that OA-GcMAF complexes could exploit the anticancer properties of both OA and GcMAF in a synergistic manner. We postulated that the stimulating effects of GcMAF on macrophages might lead to release of nitric oxide (NO). Patients with advanced cancer were treated at the Immuno Biotech Treatment Centre with OA-GcMAF-based integrative immunotherapy in combination with a low-carbohydrate, high-protein diet, fermented milk products containing naturally-produced GcMAF, Vitamin D3, omega-3 fatty acids and low-dose acetylsalicylic acid. Measuring the tumour by ultrasonographic techniques, we observed a decrease of tumour volume of about 25%. These observations demonstrate that OA, GcMAF and NO can be properly combined and specifically delivered to advanced cancer patients with significant effects on immune system stimulation and tumour volume reduction avoiding harmful side-effects.
Product Description
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Nitric oxide, 98.5%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Oleic acid, Selectophore, ≥99.0%
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material