  • Search Results
  • Favorable long-term outcomes of bilateral adrenalectomy in Cushing's disease.
Favorable long-term outcomes of bilateral adrenalectomy in Cushing's disease.
European journal of endocrinology (2014-07-01)
Andrea Oßwald, Eva Plomer, Christina Dimopoulou, Monika Milian, Rainer Blaser, Katrin Ritzel, Anne Mickisch, Ferengis Knerr, Milan Stanojevic, Klaus Hallfeldt, Jochen Schopohl, Klaus A Kuhn, Günter Stalla, Felix Beuschlein, Martin Reincke
ABSTRACT
Bilateral adrenalectomy (BADX) is an important treatment option for patients with Cushing's syndrome (CS). Our aim is to analyze the long-term outcomes, surgical, biochemical, and clinical as well as morbidity and mortality, of patients who underwent BADX. A total of 50 patients who underwent BADX since 1990 in two German centers were identified. Of them, 34 patients had Cushing's disease (CD), nine ectopic CS (ECS), and seven ACTH-independent bilateral adrenal hyperplasia (BAH). Standardized follow-up examination was performed in 36 patients with a minimum follow-up time of 6 months after BADX and a median follow-up time of 11 years. Surgical morbidity and mortality were 6 and 4% respectively. All patients were found to be in remission after BADX. Almost all Cushing's-specific comorbidities except for psychiatric diseases improved significantly. Health-related quality of life remained impaired in 45.0% of female and 16.7% of male patients compared with a healthy population. The median number of adrenal crises per 100 patient-years was four. Nelson tumor occurred in 24% of CD patients after a median time span of 51 months. Long-term mortality after 10 years was high in ECS (44%) compared with CD (3%) and BAH (14%). BADX is an effective and relatively safe treatment option especially in patients with CD. The majority of patients experience considerable improvement of Cushing's symptoms.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard