  • Search Results
  • Electron paramagnetic resonance and electron-nuclear double resonance studies of the reactions of cryogenerated hydroperoxoferric-hemoprotein intermediates.
Electron paramagnetic resonance and electron-nuclear double resonance studies of the reactions of cryogenerated hydroperoxoferric-hemoprotein intermediates.
Biochemistry (2014-07-22)
Roman Davydov, Mikhail Laryukhin, Amy Ledbetter-Rogers, Masanori Sono, John H Dawson, Brian M Hoffman
ABSTRACT
The fleeting ferric peroxo and hydroperoxo intermediates of dioxygen activation by hemoproteins can be readily trapped and characterized during cryoradiolytic reduction of ferrous hemoprotein-O2 complexes at 77 K. Previous cryoannealing studies suggested that the relaxation of cryogenerated hydroperoxoferric intermediates of myoglobin (Mb), hemoglobin, and horseradish peroxidase (HRP), either trapped directly at 77 K or generated by cryoannealing of a trapped peroxo-ferric state, proceeds through dissociation of bound H2O2 and formation of the ferric heme without formation of the ferryl porphyrin π-cation radical intermediate, compound I (Cpd I). Herein we have reinvestigated the mechanism of decays of the cryogenerated hydroperoxyferric intermediates of α- and β-chains of human hemoglobin, HRP, and chloroperoxidase (CPO). The latter two proteins are well-known to form spectroscopically detectable quasistable Cpds I. Peroxoferric intermediates are trapped during 77 K cryoreduction of oxy Mb, α-chains, and β-chains of human hemoglobin and CPO. They convert into hydroperoxoferric intermediates during annealing at temperatures above 160 K. The hydroperoxoferric intermediate of HRP is trapped directly at 77 K. All studied hydroperoxoferric intermediates decay with measurable rates at temperatures above 170 K with appreciable solvent kinetic isotope effects. The hydroperoxoferric intermediate of β-chains converts to the S = 3/2 Cpd I, which in turn decays to an electron paramagnetic resonance (EPR)-silent product at temperature above 220 K. For all the other hemoproteins studied, cryoannealing of the hydroperoxo intermediate directly yields an EPR-silent majority product. In each case, a second follow-up 77 K γ-irradiation of the annealed samples yields low-spin EPR signals characteristic of cryoreduced ferrylheme (compound II, Cpd II). This indicates that in general the hydroperoxoferric intermediates relax to Cpd I during cryoanealing at low temperatures, but when this state is not captured by reaction with a bound substrate, it is reduced to Cpd II by redox-active products of radiolysis.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)