  • Search Results
  • Abnormal lipid metabolism in collagen-induced arthritis rat model: in vitro, high resolution NMR spectroscopy based analysis.
Abnormal lipid metabolism in collagen-induced arthritis rat model: in vitro, high resolution NMR spectroscopy based analysis.
Indian journal of experimental biology (2014-07-26)
Niraj Kumar Srivastava, Shikha Sharma, Rudra N Purusottam, Neeraj Sinha, Rameshwar Singh, Deepak Sharma
ABSTRACT
Collagen-induced arthritis (CIA) was induced in female Wistar rats by intradermal injection of porcine immunization grade native collagen type II (Chondrex). Development and progression of CIA was monitored by studying histopathological, radiographical and biochemical features of arthritic manifestations in the knee joints, hind limb and blood plasma. In addition, oxidative stress status of arthritic animals was determined by measuring lipid peroxidation and the antioxidant enzymes: catalase, superoxide dismutase and glutathione peroxidase. High resolution proton NMR spectroscopy was employed for the analysis of lipid components in the lipid extracts of the joint tissue and plasma of collagen-induced arthritic and control rats. Triglyceride levels showed significant decreases in plasma (1.7 times) but were unchanged in the joint tissue of CIA rats as compared to control. One-dimensional proton NMR spectra showed a 6.2 times reduction in the quantity of choline-containing phospholipids in the plasma of CIA as compared to control rats. There was a 1.6 times elevation of choline-containing phospholipids in the joint tissue of CIA rats as compared to controls. Induction of arthritis showed a 4.0 times reduction in the level of total cholesterol in the plasma and 1.6 times elevation in the joint tissue of CIA rats as compared to controls. The ratio of saturated fatty acids to unsaturated fatty acids was 1.5 times significantly higher in joint tissue and 2.1 times significantly higher in plasma of CIA rats as compared to controls. The results demonstrated significantly altered lipid patterns in the joint tissue and plasma of collagen-induced arthritic rats as detected by one- and two-dimensional NMR spectroscopy compared with controls.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.