  • Search Results
  • Role of EZH2 protein expression in gastric carcinogenesis among Asians: a meta-analysis.
Role of EZH2 protein expression in gastric carcinogenesis among Asians: a meta-analysis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-04-08)
Lin Guo, Teng-Fei Yang, Shi-Chao Liang, Ji-Xiang Guo, Qiang Wang
ABSTRACT
The present meta-analysis aggregated the results of relevant studies to identify the role of zeste homolog 2 (EZH2) expression in gastric carcinogenesis among Asians. Related articles were found by searching the following electronic databases without language restrictions: PubMed, SpringerLink, Karger Medical and Scientific Publishers, Chinese Biomedical Database (CBM), Chinese National Knowledge Infrastructure (CNKI), and Google Scholar. Meta-analysis was performed using STATA statistical software. Crude odds ratios (ORs) or hazard ratios (HRs) with their corresponding 95 % confidence interval (95 % CI) were calculated. Ten relevant studies, which enrolled a total of 872 gastric cancer patients, were selected for statistical analysis. The most important findings of our meta-analysis was that cancer tissues exhibited higher expression levels of EZH2 protein than normal, adjacent and benign tissues (cancer tissues vs normal tissues: OR = 32.15, 95 % CI 22.58 ~ 45.79, P < 0.001; cancer tissues vs adjacent tissues: OR = 16.10, 95 % CI 11.35 ~ 22.84, P < 0.001; cancer tissues vs benign tissues: OR = 2.66, 95 % CI 1.89 ~ 3.75, P < 0.001; respectively). Furthermore, we observed positive correlations between EZH2 expression and the TNM stage (OR = 2.86, 95 % CI 1.72 ~ 4.75, P < 0.001) as well as lymph node metastasis (OR = 3.02, 95 % CI 2.01 ~ 4.53, P < 0.001) of patients with gastric carcinoma. The correlation between EZH2 expression and gastric cancer prognosis was also evaluated in the meta-analysis. Statistical analysis demonstrated that the overall survival (OS) of EZH2-negative patients was shorter than that of patients with positive expressions of EZH2 (HR = 0.54, 95 % CI = 0.05 ~ 1.03, P = 0.032). Our meta-analysis confirmed the view that EZH2 expression might participate in the development of gastric carcinogenesis. Thus, EZH2 protein may be a valuable biomarker for the diagnosis and prognosis of gastric cancer.
Sigma-Aldrich
Anti-EZH2 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution