  • Search Results
  • FoxO3a is an antimelanogenic factor that mediates antioxidant-induced depigmentation.
FoxO3a is an antimelanogenic factor that mediates antioxidant-induced depigmentation.
The Journal of investigative dermatology (2013-11-29)
Juewon Kim, Hyunjung Choi, Eun-Gyung Cho, Tae R Lee
ABSTRACT
Forkhead box-O (FoxO) family transcriptional factors control the expression of many genes involved in a variety of cellular processes. Melanogenesis is an oxidizing process; therefore, many antioxidants are used to inhibit melanin production. However, their mechanism of action is poorly understood. In this study, we investigated the role of FoxO3a, which is a key factor in oxidative stress-related cellular responses in melanogenesis. When FoxO3a expression was inhibited, the expression of melanogenic genes and melanin levels increased. In contrast, the overexpression of wild-type FoxO3a and the increased nuclear translocation induced by the phosphoinositide 3-kinase inhibitors or by Akt inhibition reversed these phenomena. This effect was not observed when FoxO3a harbored a deletion in the nuclear localization signal, indicating that its nuclear translocation is important for the regulation of melanogenesis. When antioxidants such as vitamin C, N-acetylcysteine, and Trolox were applied to MNT1 cells, melanin levels decreased in parallel with FoxO3a nuclear translocation, and this effect disappeared with FoxO3a-directed small interfering RNA treatment. Because FoxO3a orchestrates the expression of many genes in order to regulate cellular phenotypes in a variety of environmental states, this gene, a factor involved in melanogenesis regulation, may represent a good target for studying antimelanogenic signaling pathways and for designing pharmacological or antimelanogenic agents that regulate melanin synthesis.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.