  • Search Results
  • The rat androgen-binding protein (ABP/SHBG) gene contains triplet repeats similar to unstable triplets: evidence that the ABP/SHBG and the fragile X-related 2 genes overlap.
The rat androgen-binding protein (ABP/SHBG) gene contains triplet repeats similar to unstable triplets: evidence that the ABP/SHBG and the fragile X-related 2 genes overlap.
Steroids (1998-01-23)
D R Joseph
ABSTRACT
The rat androgen-binding protein/sex hormone-binding globulin (ABP/SHBG) gene is regulated by promoters P1 and PA. P1 regulates the mRNA encoding secreted ABP/SHBG, whereas PA regulates an alternate mRNA which encodes a modified protein that is targeted to the nucleus. Promoter PA is GC rich, consisting of 70-80% GC residues. During routine BLAST sequence analysis it was discovered that this GC-rich region is highly related to the human fragile X-related protein 2 (FXR-2) 5'-untranslated RNA sequence. Furthermore, the nucleotide coding sequence of the initial 14 FXR-2 amino acid residues was identical in the ABP/SHBG gene. The 5'-untranslated FXR-2 sequence contains triplet (CGG) repeats, which are also present in the rat ABP/SHBG gene. The meiotic instability of CGG repeats in the human fragile X (FMR1) gene causes the fragile X mental retardation syndrome. The data presented here suggest that the ABP/SHBG and FXR-2 genes overlap with each gene transcribed in the opposite direction. In support of this structure, the human ABP/SHBG and the FXR-2 genes map to the same site on chromosome 17. Thus, the ABP/SHBG gene contains triplet repeats in the alternate promoter PA. It will be of particular interest to determine if triplet instability affects ABP/SHBG gene expression. A triplet instability in the X-linked androgen receptor gene causes spinal and bulbar muscular atrophy.
Sigma-Aldrich
Anti-FXR2 antibody, Mouse monoclonal, clone A42, purified from hybridoma cell culture
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.