  • Search Results
  • Dietary selenomethionine exposure alters swimming performance, metabolic capacity and energy homeostasis in juvenile fathead minnow.
Dietary selenomethionine exposure alters swimming performance, metabolic capacity and energy homeostasis in juvenile fathead minnow.
Aquatic toxicology (Amsterdam, Netherlands) (2014-07-08)
D Landon McPhee, David M Janz
ABSTRACT
Selenium (Se) is known to cause chronic toxicity in aquatic species. In particular, dietary exposure of fish to selenomethionine (SeMet), the primary form of Se in the diet, is of concern. Recent studies suggest that chronic exposure to elevated dietary SeMet alters energy and endocrine homeostasis in adult fish. However, little is known about the direct effects of dietary SeMet exposure in juvenile fish. The objective of the present study was to investigate sublethal physiological effects of dietary SeMet exposure in juvenile fathead minnow (Pimephales promelas). Twenty days-post-hatch fathead minnow were exposed for 60 days to different measured concentrations (2.8, 5.4, 9.9, 26.5 μg Se/g dry mass [dm]) of Se in food in the form of SeMet. After exposure, samples were collected for Se analysis and fish were subjected to a swimming performance challenge to assess critical swim speed (Ucrit), tail beat frequency and tail beat amplitude, oxygen consumption (MO2), cost of transport (COT), standard metabolic rate (SMR), active metabolic rate (AMR), and factorial aerobic scope (F-AS). Ucrit was decreased in the 26.5 μg Se/g dm exposure group compared to the control group. Tail beat frequency and tail beat amplitude were significantly reduced in fish fed 9.9 and 26.5 μg Se/g. An increase in MO2 and COT was observed in the 9.9 and 26.5 μg Se/g exposure groups compared to the control group. While the AMR of the high dose group was increased relative to control, there were no significant differences in SMR and F-AS. Energy storage capacity was measured via whole body triglyceride and glycogen concentrations. Triglyceride concentrations in non-swam fish were elevated in the 5.4 μg Se/g group relative to controls. Fatigued (swam) fish had significantly lower whole body triglycerides than non-swam fish. All non-swam SeMet exposure groups had significantly decreased whole body glycogen concentrations compared to controls, while the 5.4 and 26.5 μg Se/g exposure groups had significantly greater whole body glycogen concentrations in swam versus non-swam fish. A decrease in whole body cortisol was observed in swam fish in the 5.4 μg Se/g exposure group compared to control fish. Whole body cortisol was greater in control, 9.9 and 26.5 μg Se/g swam fish compared to non-swam fish. These results suggest that exposure to environmentally relevant concentrations of dietary SeMet impairs swimming performance, aerobic capacity, and energy homeostasis, potentially impacting survivability of juvenile fish in Se impacted aquatic ecosystems.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Glycogen from bovine liver, ≥85%
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), for DNA precipitations
Sigma-Aldrich
Glycogen from oyster, ≥75% dry basis
Sigma-Aldrich
Seleno-L-methionine, ≥98% (TLC), powder
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), ≥85%
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Seleno-DL-methionine, ≥99% (TLC)
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Glycogen from oyster, Type XI