  • Search Results
  • Seedling emergence, growth and trace elements tolerance and accumulation by Lamiaceae species in a mine soil.
Seedling emergence, growth and trace elements tolerance and accumulation by Lamiaceae species in a mine soil.
Chemosphere (2014-07-30)
A Parra, R Zornoza, E Conesa, M D Gómez-López, A Faz
ABSTRACT
The potential use of three Laminaceae species (Lavandula dentata, Rosmarinus officinalis and Thymus vulgaris) for the phytostabilisation of a trace elements contaminated (acid) soil has been evaluated. These species were grown in mine tailing soil unamended (TS) and amended with calcium carbonate and pig manure (ATS), and unpolluted substrate for control (CT); plant growth, root characterisation, soil trace elements contents and their accumulation in plants were measured. Results indicated that seed emergence was independent from substrate characteristics, but seedlings died in TS with 40% survival in ATS. The biomass of L. dentata and T. vulgaris and root development in R. officinalis were negatively affected when grown in TS but without differences between ATS and CT. Applicating amendments reduced soil exchangeable and extractable fractions concentrations of trace elements in ATS compared with TS. The establishment of L. dentata and R. officinalis were related to trace elements immobilisation. Trace element concentrations in plants grown in tailing soils were similar to those reported for control, although applicating amendments reduced Zn accumulation in all species, and favoured increased absorption and aerial translocation of As and Pb by L. dentata and T. vulgaris; nonetheless, levels were below toxicity thresholds. Thus, these species fulfill the criteria for phytostabilisation purposes, aided by employing amendments.
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%