  • Search Results
  • Molecular responses in digestive tract of juvenile common carp after chronic exposure to sublethal tributyltin.
Molecular responses in digestive tract of juvenile common carp after chronic exposure to sublethal tributyltin.
Ecotoxicology and environmental safety (2014-08-19)
Zhi-Hua Li, Ping Li, Ze-Chao Shi
ABSTRACT
The effect of long-term exposure to tributyltin (TBT) on the intestine-related biochemical biomarkers in common carp was investigated in this study. Fish were exposed at sub-lethal concentrations of TBT (75 ng/L, 0.75 and 7.5 μg/L) for 60 days. Multiple biomarkers were measured, including digestive enzymes (trypsin, lipase and amylase), antioxidant responses (malondialdehyde (MDA) and total antioxidative capacity (T-AOC)), RNA/DNA ratio and the expression of digestive-related genes (try, lipc and amy). TBT exposure at 0.75 and 7.5 μg/L led to significantly inhibited activities of all digestive enzymes. At higher concentration of TBT, oxidative stress was apparent as reflected by the significant higher MDA content in the fish intestine, associated with an inhibition of T-AOC activities. After 60 days, the RNA/DNA ratio in fish intestine was significantly lower in groups exposed to TBT at higher concentrations (0.75 and 7.5 μg/L). In addition, the expression levels of try, lipc and amy in intestine of all treated fish were inhibited, even at the environmental concentration (75 ng/L). Our results suggest that long-term exposure to TBT could result in different responses of intestine-related biochemical biomarkers in fish, which could be used as new potential indicators for monitoring residual TBT present in aquatic environment.
Product Description
Ribonucleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid from baker's yeast (S. cerevisiae)
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta
Sigma-Aldrich
Tributyltin hydride solution, 1 M in cyclohexane
Sigma-Aldrich
Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid, transfer, phenylalanine specific from brewer's yeast
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
Ribonucleic acid from calf liver, Type IV
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization