  • Search Results
  • Hypocholesterolaemic pharmaceutical simvastatin disrupts reproduction and population growth of the amphipod Gammarus locusta at the ng/L range.
Hypocholesterolaemic pharmaceutical simvastatin disrupts reproduction and population growth of the amphipod Gammarus locusta at the ng/L range.
Aquatic toxicology (Amsterdam, Netherlands) (2014-08-05)
Teresa Neuparth, Carla Martins, Carmen B de Los Santos, Maria H Costa, Irene Martins, Pedro M Costa, Miguel M Santos
ABSTRACT
Simvastatin (SIM), a hypocholesterolaemic drug, is among the most widely used pharmaceuticals worldwide and is therefore of emerging environmental concern. Despite the ubiquitous nature of SIM in the aquatic ecosystems, significant uncertainties exist about sublethal effects of the drug in aquatic organisms. Therefore, here we aimed at investigating a multi-level biological response in the model amphipod Gammarus locusta, following chronic exposures to low levels of SIM (64 ng/L to 8 μg/L). The work integrated a battery of key endpoints at individual-level (survival, growth and reproduction) with histopathological biomarkers in hepatopancreas and gonads. Additionally, an individual-based population modelling was used to project the ecological costs associated with long-term exposure to SIM at the population level. SIM severely impacted growth, reproduction and gonad maturation of G. locusta, concomitantly to changes at the histological level. Among all analysed endpoints, reproduction was particularly sensitive to SIM with significant impact at 320 ng/L. These findings have important implications for environmental risk assessment and disclose new concerns about the effects of SIM in aquatic ecosystems.
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Simvastatin, analytical standard
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material