MilliporeSigma
Syncope in genotype-negative long QT syndrome family members.
The American journal of cardiology (2014-09-01)
Louise R A Olde Nordkamp, Martin H Ruwald, Ilan Goldenberg, Wouter Wieling, Scott McNitt, Bronislava Polonsky, Arthur A M Wilde, Nynke van Dijk, Arthur J Moss
ABSTRACT
Unaffected long-QT syndrome family members (FMs) frequently experience syncope. The aims of this study were to test the hypothesis that syncope events in FMs are benign events and to compare clinical characteristics, triggers eliciting the syncope events, and long-term outcomes between FMs and those with LQT1 or LQT2 mutations from the international Long QT Syndrome Registry. A total of 679 FMs, 864 LQT1 patients, and 782 LQT2 patients were included. Seventy-eight FMs (11%) experienced cardiovascular events. Almost all cardiovascular events were nonfatal syncope; only 1 FM, with an additional mitral valve prolapse, experienced aborted cardiac arrest during exercise. The mean age at first syncope in FMs was 17 years, and female FMs experienced syncope more frequently than male FMs (14% vs 9%, p = 0.027). Syncope was more frequently triggered by exercise in LQT1 patients (43% in LQT1 patients vs 5% in FMs, p <0.001), while syncope triggered by a variety of other triggers was more frequent in FMs (54% in FMs vs 22% in LQT1 patients and 30% in LQT2 patients, p <0.001 for both). None of the FMs experienced aborted cardiac arrest or sudden cardiac death after the first syncopal episode. In conclusion, syncope is frequently present in FMs, and these syncopal events occurred more frequently in female than in male FMs, with an increased incidence in midadolescence. Triggers eliciting the syncopal events were different between FMs and patients with long-QT syndrome mutations. Hence, the type of trigger is useful in distinguishing between high- and low-risk syncope. These data indicate that FMs from families with LQTS have a benign form of syncope, most likely related to vasovagal syncope and not ventricular tachyarrhythmic syncope.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta