  • Search Results
  • Time-dependent degenerative transformations in the lipidome of chalazia.
Time-dependent degenerative transformations in the lipidome of chalazia.
Experimental eye research (2014-08-26)
Jadwiga C Wojtowicz, Igor A Butovich, Anne McMahon, Robert N Hogan, Kamel M Itani, Ronald Mancini, Mike Molai, Emily Linsenbardt
ABSTRACT
The aim of this prospective study was to conduct histopathologic and lipidomic analyses of chalazia, in order to evaluate time-dependent changes in the lesion. Samples of surgically excised chalazia were collected over a period of 12 months from 10 patients (mean age 41 years; range, 23-58) with clinically diagnosed chalazia, who underwent scheduled surgery. The ages of chalazia varied from 2 to 28 weeks. To confirm the clinical diagnoses, the morphology of collected tissue samples was evaluated histologically after hematoxylin and eosin staining. The lipids from individual chalazia were analyzed by high-performance liquid chromatography-mass spectrometry and compared with authentic lipid standards and with the lipids of meibum collected from normal controls. We observed gradual, lesion age-dependent transformation of the lipidome of chalazia from an almost normal meibum-like composition to a very different kind of lipidome. A rapid initial increase in the free cholesterol content was followed by a gradual replacement of extremely long chain meibomian-type lipids with a mixture of shorter-chain cholesteryl esters of the C14-C18 family, triacylglycerols, ceramides, phospholipids and sphingomyelins. In addition, a rapid disappearance of wax esters and cholesteryl esters of (1-O)-acyl-omega-hydroxy fatty acids from the lipidome of aging chalazia was observed. Our results are indicative of dramatic, time-dependent changes in the lesion that may involve cholesterol as a trigger and/or a marker of subsequent degeneration of the meibomian lipidome. We hypothesize that early inhibition of these transformations may be useful in reversing the course of the disease.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG