Product Description
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.025mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.006mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.003mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.001mm, temporary acrylic support, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.015mm, 99.99+%
Supelco
Lead (impurities), manufactured by MBH Analytical Ltd (83X PR8)
Sigma-Aldrich
Lead, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Lead, foil, light tested, 50x50mm, thickness 0.0125mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Lead, foil, 1m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 10mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 75x100mm, thickness 2.7mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.125mm, as rolled, 99.95%