  • Search Results
  • A prospective, randomized, multicenter trial of Surgisis Gold, a biologic prosthetic, as a sublay reinforcement of the fascial closure after open bariatric surgery.
A prospective, randomized, multicenter trial of Surgisis Gold, a biologic prosthetic, as a sublay reinforcement of the fascial closure after open bariatric surgery.
Surgery (2014-09-23)
Michael G Sarr, Neil E Hutcher, Scott Snyder, Jason Hodde, Brennan Carmody
ABSTRACT
This prospective, multicenter, randomized clinical trial was performed to compare the effectiveness of the bioprosthesis Surgisis Gold (Cook Biotech, Inc, West Lafayette, IN) to suture closure alone in reinforcing the abdominal wall after open Roux-en-Y gastric bypass (RYGB). A total of 402 adult patients (body mass index ≥35 kg/m(2)) scheduled to undergo open RYGB procedures were randomized to receive either Surgisis Gold placed as a sublay directly under and opposed to the posterior rectus fascia without intervening preperitoneal fat or suture closure alone. Peri- and postoperative complications were tracked. Incisional hernia formation in each group was examined at 6 weeks, 3, 6, and 9 months, and 1 and 2 years after RYGB. A total of 380 (78 male; 302 female) patients with a mean age of 45 years (range 18-69) and a mean BMI of 48 (range 35-79) underwent RYGB. There were no differences in patient demographics or comorbidities between groups. Placement of the Surgisis Gold increased operating time by an average of 14 minutes. Complications were more common in the Surgisis Gold group with significantly more wound events and seroma formation compared with the suture closure alone group. At final follow-up, 32 of 185 (17%) of patients in the reinforced arm and 38 of 195 (20%) in the suture alone arm developed an incisional hernia (P = .6). In this challenging patient population, the use of Surgisis Gold for reinforcing the abdominal wall after open RYGB was not shown to be greatly different from a primary suture repair.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I (Sigma Type VII), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.