  • Search Results
  • Pre-emptive analgesia with pregabalin and celecoxib decreases postsurgical pain following maxillomandibular advancement surgery: a randomized controlled clinical trial.
Pre-emptive analgesia with pregabalin and celecoxib decreases postsurgical pain following maxillomandibular advancement surgery: a randomized controlled clinical trial.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-09-23)
Joseph E Cillo, David J Dattilo
ABSTRACT
The purpose of this study was to determine the efficacy of pre-emptive analgesia with pregabalin and celecoxib on narcotic consumption and perceived pain in adult patients undergoing maxillomandibular advancement surgery for obstructive sleep apnea. This was a prospective, randomized, double-blinded, placebo-controlled study of adult patients undergoing elective maxillomandibular advancement surgery for obstructive sleep apnea. The groups received a masked 1-time preoperative oral dose of pregabalin 150 mg and celecoxib 400 mg (experimental group) or lactose powder 2 g (placebo group). In the postoperative period, pain management consisted of intravenous morphine patient-controlled analgesia and oral oxycodone 5 mg and acetaminophen 325 mg. Patients completed a daily pain and narcotic log. Statistical significance between group means was determined by the 2-tailed independent t test. There were statistically significant differences between the pregabalin plus celecoxib and placebo groups in average intravenous morphine consumption per 4-hour interval (6.0 ± 5.9 vs 9.3 ± 7.9 mg; P < .05), mean daily narcotic pill consumption (2.9 ± 2.9 vs 6.8 ± 1.8 pills; P < .05), and mean daily visual analog scale scores (4.3 ± 3.5 vs 5.5 ± 5.0; P < .05). Within the limitations of this study, a 1-time preoperative oral dose of pregabalin and celecoxib before adult maxillomandibular advancement surgery for obstructive sleep apnea decreased mean intravenous morphine consumption, mean daily narcotic pill consumption, and mean patient perceived pain.
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Oxycodone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
Sigma-Aldrich
Morphine sulfate salt pentahydrate
Supelco
Oxycodone hydrochloride, analytical standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®