MilliporeSigma
Selective uncaging of DNA through reaction rate selectivity.
Organic letters (2014-09-19)
Alexandre Rodrigues-Correia, Diana Knapp-Bühle, Joachim W Engels, Alexander Heckel
ABSTRACT
The synthesis and use of the new nucleobase-caged nucleotides dT(pHP) and dT(NDEACM) is reported. Through a combination of time and wavelength selectivity four levels of selective uncaging with only two cages, and only two wavelengths, were obtained. The new residue dT(pHP) can be uncaged at 313 nm without the formation of unwanted cyclic pyridine dimers.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus), Type XI, Highly polymerized
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization