  • Search Results
  • The inhibitor of differentiation isoform Id1b, generated by alternative splicing, maintains cell quiescence and confers self-renewal and cancer stem cell-like properties.
The inhibitor of differentiation isoform Id1b, generated by alternative splicing, maintains cell quiescence and confers self-renewal and cancer stem cell-like properties.
Cancer letters (2014-12-03)
Irene Manrique, Paul Nguewa, Anne-Marie Bleau, Estanislao Nistal-Villan, Ines Lopez, Maria Villalba, Ignacio Gil-Bazo, Alfonso Calvo
ABSTRACT
Id1 has been shown to play a critical role in tumorigenesis and angiogenesis. Moreover, recent reports have involved Id1 in the maintenance of cancer stem cell features in some tumor types. The Id1 gene generates two isoforms through alternative splicing: Id1a and Id1b. We have investigated the role of each isoform in cancer development. Using lentiviral systems we modified the endogenous expression of each of these isoforms in cancer cells and analyzed their biological effect both in vitro and in vivo. Overexpression of Id1b in murine CT26 and 3LL cells caused a G0/G1 cell cycle arrest and reduced proliferation, clonogenicity and phospho-ERK1/2 levels, while increasing p27 levels. High levels of Id1a had an opposite effect and the proportion of cells in the S phase increased significantly. In vivo models confirmed the inhibitory role of Id1b in primary tumor growth and metastasis. Through microarray analysis we found that the cancer stem cell (CSC) markers ALDH1A1 and Notch-1 were up-regulated specifically in Id1b-overexpressing cells. By using qPCR we also found overexpression of Sca-1, Tert, Sox-2 and Oct-4 in these cells. Increased levels of Id1b promoted self-renewal and CSC-like properties, as shown by their high capacity for developing secondary tumorspheres and retaining the PKH26 dye. The acquisition of CSC phenotype was confirmed in human PC-3 cells that overexpressed Id1b. Our results show that Id1b maintains cells in a quiescent state and promotes self-renewal and CSC-like features. On the contrary, Id1a promotes cell proliferation.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard