  • Search Results
  • Distinct antifibrogenic effects of erlotinib, sunitinib and sorafenib on rat pancreatic stellate cells.
Distinct antifibrogenic effects of erlotinib, sunitinib and sorafenib on rat pancreatic stellate cells.
World journal of gastroenterology (2014-07-01)
Anne Elsner, Falko Lange, Brit Fitzner, Martin Heuschkel, Bernd Joachim Krause, Robert Jaster
ABSTRACT
To study if three clinically available small molecule kinase inhibitors (SMI), erlotinib, sunitinib and sorafenib, exert antifibrogenic effects on pancreatic stellate cells (PSC) and analyze the basis of their action. Cultured rat PSC were exposed to SMI. Cell proliferation and viability were assessed employing 5-bromo-2'-deoxyuridine incorporation assay and flow cytometry, respectively. 2-Deoxy-2-[(18)F] fluoroglucose ((18)F-FDG) uptake was measured to study metabolic activity. Exhibition of the myofibroblastic PSC phenotype was monitored by immunofluorescence analysis of α-smooth muscle actin (α-SMA) expression. Levels of mRNA were determined by real-time PCR, while protein expression and phosphorylation were analyzed by immunoblotting. Transforming growth factor-β1 (TGF-β1) levels in culture supernatants were quantified by ELISA. All three SMI inhibited cell proliferation and (18)F-FDG uptake in a dose-dependent manner and without significant cytotoxic effects. Furthermore, additive effects of the drugs were observed. Immunoblot analysis showed that sorafenib and sunitib, but not erlotinib, efficiently blocked activation of the AKT pathway, while all three drugs displayed little effect on phosphorylation of ERK1/2. Cells treated with sorafenib or sunitinib expressed less interleukin-6 mRNA as well as less collagen type 1 mRNA and protein. Sorafenib was the only drug that also upregulated the expression of matrix metalloproteinase-2 and reduced the secretion of TGF-β1 protein. All three drugs showed insignificant or discordant effects on the mRNA and protein levels of α-SMA. The tested SMI, especially sorafenib, exert inhibitory effects on activated PSC, which should be further evaluated in preclinical studies.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody